missing translation for 'onlineSavingsMsg'
Learn More

(R,E)-5-([1,1-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoic Acid, TRC

Product Code. 30467541
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467541 5 g 5g
30468847 100 mg 100mg
30465427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30467541 Supplier Toronto Research Chemicals Supplier No. TRCM3452055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1012341-48-8
Molecular Formula C23H27NO4
Molecular Weight (g/mol) 381.47
Synonym Sacubitril Impurity 22
IUPAC Name (E,4R)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pent-2-enoic acid
SMILES C\C(=C/[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)OC(C)(C)C)\C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C23H27NO4/c1-16(21(25)26)14-20(24-22(27)28-23(2,3)4)15-17-10-12-19(13-11-17)18-8-6-5-7-9-18/h5-14,20H,15H2,1-4H3,(H,24,27)(H,25,26)/b16-14+/t20-/m0/s1
Formula Weight 381.194
Chemical Name or Material (R,E)-5-([1,1-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.