missing translation for 'onlineSavingsMsg'
Learn More

(±)-4-Methyl-1-nonanol, TRC

Product Code. 30467543
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467543 100 mg 100mg
30467551 50 mg 50mg
30471821 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30467543 Supplier Toronto Research Chemicals Supplier No. TRCM385535100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-47-0
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
Synonym 4-Methyl-1-nonanol
IUPAC Name 4-methylnonan-1-ol
SMILES CCCCCC(C)CCCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H22O/c1-3-4-5-7-10(2)8-6-9-11/h10-11H,3-9H2,1-2H3
Formula Weight 158.1671
Chemical Name or Material (±)-4-Methyl-1-nonanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.