missing translation for 'onlineSavingsMsg'
Learn More

4’-Methylbiphenyl-2-carboxylic Acid, TRC

Product Code. 30468591
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468591 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468591 Supplier Toronto Research Chemicals Supplier No. TRCM29412025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Methylbiphenyl-2-carboxylic Acid
CAS 7148-03-0
Molecular Formula C14 H12 O2
Quantity 25 g
Synonym 4'-Methyl-[1,1'-biphenyl]-2-carboxylic Acid, 4'-Methyl-2-biphenylcarboxylic Acid, 2-(4-Methylphenyl)benzoic Acid, 2-(p-Tolyl)benzoic Acid, 2-Carboxy-4'-methylbiphenyl, 4-Methyl-2'-biphenylcarboxylic Acid, 4'-Methyl-2-biphenylcarboxylic Acid, 4'-Methyl-2-biphenylylcarboxylic Acid, 4'-Methyl[1,1'-biphenyl]-2-carboxylic Acid, NSC 26069
InChI Formula InChI=1S/C14H12O2/c1-10-6-8-11(9-7-10)12-4-2-3-5-13(12)14(15)16/h2-9H,1H3,(H,15,16)
SMILES Cc1ccc(cc1)c2ccccc2C(=O)O
IUPAC Name 2-(4-methylphenyl)benzoic acid
Molecular Weight (g/mol) 212.244
Formula Weight 212.084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.