missing translation for 'onlineSavingsMsg'
Learn More

rac-gamma-Nonanolactone, TRC

Product Code. 30469226
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30469226 500 g
30467997 50 g
30475477 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469226 Supplier Toronto Research Chemicals Supplier No. TRCN649360500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-61-0
Molecular Formula C9 H16 O2
Molecular Weight (g/mol) 156.22
Synonym 4-Nonanolide, 4-Pentyl-butanolide, γ-Amylbutyrolactone, γ-Nonanolide, Aldehyde C-18, Apricolin, Cocos aldehyde, Dihydro-5-pentyl-2(3H)-furanone, Prunolide
IUPAC Name 5-pentyloxolan-2-one
SMILES CCCCCC1CCC(=O)O1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C9H16O2/c1-2-3-4-5-8-6-7-9(10)11-8/h8H,2-7H2,1H3
Formula Weight 156.115
Chemical Name or Material gamma-Nonalactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.