missing translation for 'onlineSavingsMsg'
Learn More

Monononyl Phthalate, TRC

Product Code. 30472051
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472051 25 mg 25mg
30471237 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30472051 Supplier Toronto Research Chemicals Supplier No. TRCM56660025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24539-59-1
Molecular Formula C17 H24 O4
Molecular Weight (g/mol) 292.37
Synonym 1,2-Benzenedicarboxylic Acid Monononyl Ester, Nonyl Alcohol Phthalate, Phthalic Acid Monononyl Ester, Mono(n-nonyl) Phthalate
IUPAC Name 2-nonoxycarbonylbenzoic acid
SMILES CCCCCCCCCOC(=O)c1ccccc1C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H24O4/c1-2-3-4-5-6-7-10-13-21-17(20)15-12-9-8-11-14(15)16(18)19/h8-9,11-12H,2-7,10,13H2,1H3,(H,18,19)
Formula Weight 292.167
Chemical Name or Material Monononyl Phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.