missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mefenamic-d4 Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Mefenamic-d4 Acid |
| CAS | 1216745-79-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 D4 H11 N O2 |
| Quantity | 10 mg |
| Synonym | Benzoic-2,3,4,5-d4 acid, 6-[(2,3-dimethylphenyl)amino]- (ACI), 6-[(2,3-Dimethylphenyl)amino]benzoic-2,3,4,5-d4 acid (ACI), Mefenamic-d4 Acid, Mefenamic Acid D4 (benzoic acid 3,4,5,6-D4) |
| InChI Formula | InChI=1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)/i3D,4D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)O)c(Nc2cccc(C)c2C)c1[2H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(2,3-dimethylanilino)benzoic acid |
| Molecular Weight (g/mol) | 245.3097 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?