missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline Hydrochloride, TRC

Product Code. 30480519
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480519 5 g 5g
30470487 1 g 1g
30484031 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480519 Supplier Toronto Research Chemicals Supplier No. TRCM2883085G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2739-12-0
Molecular Formula C7H9N . HCl
Molecular Weight (g/mol) 143.61
Synonym N-Methylanilinium Chloride, N-Methylbenzenamine Hydrochloride, Anilinomethane Hydrochloride
IUPAC Name N-methylaniline;hydrochloride
SMILES Cl.CNc1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H9N.ClH/c1-8-7-5-3-2-4-6-7;/h2-6,8H,1H3;1H
Formula Weight 143.0502
Chemical Name or Material N-Methylaniline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.