missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide 3-Isomer, TRC

Product Code. 30481032
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481032 50 mg 50mg
30476697 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481032 Supplier Toronto Research Chemicals Supplier No. TRCL32276550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61643-23-0
Molecular Formula C12 H9 F3 N2 O2
Molecular Weight (g/mol) 270.21
Synonym 5-Methyl-N-[3-(trifluoromethyl)phenyl]isoxazole-4-carboxamide, 4-Isoxazolecarboxamide, 5-methyl-N-[3-(trifluoromethyl)phenyl]- (9CI), Leflunomide 3-Isomer, N-(3'-Trifluoromethylphenyl)-5-methylisoxazole-4-carboxamide
IUPAC Name 5-methyl-N-[3-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
SMILES Cc1oncc1C(=O)Nc2cccc(c2)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H9F3N2O2/c1-7-10(6-16-19-7)11(18)17-9-4-2-3-8(5-9)12(13,14)15/h2-6H,1H3,(H,17,18)
Formula Weight 270.0616
Chemical Name or Material 5-Methyl-N-[3-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.