missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetone, TRC

Product Code. 30481588
Change view
Click to view available options
Quantity:
1 g
50 g
Unit Size:
1g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481588 50 g 50g
30475523 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481588 Supplier Toronto Research Chemicals Supplier No. TRCM26093050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methoxyphenylacetone
CAS 3027-13-2
Molecular Formula C10 H12 O2
Quantity 50 g
Synonym 2-Propanone, 1-(3-methoxyphenyl)-, 2-Propanone, (m-methoxyphenyl)- (6CI,7CI), 2-Propanone, 1-(m-methoxyphenyl)- (8CI), 1-(3-Methoxyphenyl)-2-propanone, (3-Methoxyphenyl)acetone, (m-Methoxyphenyl)acetone, 1-(3-Methoxyphenyl)acetone, 1-(m-Methoxyphenyl)-2-propanone, 3-Methoxybenzyl methyl ketone, 3'-Methoxyphenyl-2-propanone, m-Methoxybenzyl methyl ketone, m-Methoxyphenylacetone, meta-Methoxyphenylacetone
InChI Formula InChI=1S/C10H12O2/c1-8(11)6-9-4-3-5-10(7-9)12-2/h3-5,7H,6H2,1-2H3
SMILES COc1cccc(CC(=O)C)c1
IUPAC Name 1-(3-methoxyphenyl)propan-2-one
Molecular Weight (g/mol) 164.2011
Formula Weight 164.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.