missing translation for 'onlineSavingsMsg'
Learn More

Methyl Flufenpyr, TRC

Product Code. 30481787
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481787 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481787 Supplier Toronto Research Chemicals Supplier No. TRCM633840250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Flufenpyr
CAS 188489-52-3
Molecular Formula C15H11ClF4N2O4
Quantity 250 mg
Synonym Methyl 2-[2-Chloro-4-fluoro-5-[5-methyl-6-oxo-4-(trifluoromethyl)-1(6H)-pyridazinyl]phenoxy]acetate
InChI Formula InChI=1S/C15H11ClF4N2O4/c1-7-8(15(18,19)20)5-21-22(14(7)24)11-4-12(9(16)3-10(11)17)26-6-13(23)25-2/h3-5H,6H2,1-2H3
SMILES COC(=O)COc1cc(N2N=CC(=C(C)C2=O)C(F)(F)F)c(F)cc1Cl
IUPAC Name methyl 2-[2-chloro-4-fluoro-5-[5-methyl-6-oxo-4-(trifluoromethyl)pyridazin-1-yl]phenoxy]acetate
Molecular Weight (g/mol) 394.71
Formula Weight 394.0343
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.