missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Methoxy-2-oxoethyl)sulfanyl]acetic Acid, TRC

Product Code. 30482422
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482422 100 mg 100mg
30464416 50 mg 50mg
30483742 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482422 Supplier Toronto Research Chemicals Supplier No. TRCM486110100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91114-48-6
Molecular Formula C5H8O4S
Molecular Weight (g/mol) 164.18
IUPAC Name 2-(2-methoxy-2-oxoethyl)sulfanylacetic acid
SMILES COC(=O)CSCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H8O4S/c1-9-5(8)3-10-2-4(6)7/h2-3H2,1H3,(H,6,7)
Formula Weight 164.0143
Chemical Name or Material 2-[(2-Methoxy-2-oxoethyl)sulfanyl]acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.