missing translation for 'onlineSavingsMsg'
Learn More

NG-Monomethyl-L-arginine Acetate, TRC

Product Code. 30483214
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483214 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483214 Supplier Toronto Research Chemicals Supplier No. TRCM56500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NG-Monomethyl-L-arginine Acetate
CAS 53308-83-1
Purity Grade Notes HPLC
Molecular Formula C7 H16 N4 O2 . C2 H4 O2
Quantity 25 mg
Synonym N5-[Imino(methylamino)methyl]-L-ornithine Acetate, L-NMMA, ANO 1020, Tilarginine Acetate
InChI Formula InChI=1S/C7H16N4O2.C2H4O2/c1-10-7(9)11-4-2-3-5(8)6(12)13;1-2(3)4/h5H,2-4,8H2,1H3,(H,12,13)(H3,9,10,11);1H3,(H,3,4)/t5-;/m0./s1
SMILES CNC(=N)NCCC[C@H](N)C(=O)O.CC(=O)O
IUPAC Name acetic acid;(2S)-2-amino-5-[(N-methylcarbamimidoyl)amino]pentanoic acid
Molecular Weight (g/mol) 248.28
Formula Weight 248.1485
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.