missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 12-Hydroxystearate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 12-hydroxystearate |
| CAS | 141-23-1 |
| Molecular Formula | C19 H38 O3 |
| Quantity | 500 mg |
| Synonym | 12-Hydroxyoctadecanoic acid methyl ester, Methyl 12-hydroxyoctadecanoate, Methyl 12-hydroxystearate, 12-Hydroxyoctadecanoic acid, methyl ester |
| InChI Formula | InChI=1S/C19H38O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h18,20H,3-17H2,1-2H3 |
| SMILES | CCCCCCC(O)CCCCCCCCCCC(=O)OC |
| IUPAC Name | methyl 12-hydroxyoctadecanoate |
| Molecular Weight (g/mol) | 314.5 |
| Formula Weight | 314.2821 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?