Description
- Wide range of precise and accurate flow rates for isocratic or gradient HPLC
- Provides high-efficiency mixing for accurate and repeatable gradients over a wide flow rate range
- Powerful, smooth flow rates from a robust and laboratory-proven pump design
- Enables simple and easy upgrades from isocratic to binary, tertiary, or quaternary gradients
Specifications
Specifications
| Max. Flow Rate | 200 mL/min. |
| Min. Flow Rate | 10 μL/min |
| Voltage | 110/220 V |
| Product Type | HPLC Pump |