missing translation for 'onlineSavingsMsg'
Learn More

AseptiQuik X Body 1in Hose Barb

Brand:  CPC Colder Products Company AQX17016

AseptiQuik X Body 1in Hose Barb

Product Code. 30968698

  • ONLINE EXCLUSIVE PRICE
    99.90 €
    97.44 € / Pack of 1
    Save 2.46 €
    2% Off
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.