missing translation for 'onlineSavingsMsg'
Learn More

Anritsu™ 30mm Pipe Silicone Cable Surface Probe

Product Code. 15308714
Change view
Click to view available options
Length (Metric) Cable:
1 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Unit Size
15308714 1 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15308714 Supplier Anritsu™ Supplier No. A231E001TC1ASP
  • ONLINE EXCLUSIVE PRICE
    202.65 €
    127.39 € / Pack of 1
    Save 75.26 €
    37% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.