missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 32 oz. Narrow Mouth Bottle

Product Code. 17140056
Click to view available options
Color:
Blue
Gray
Seafoam
Trout Green
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17140056

Brand: Nalgene 20782027

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All the effectiveness of the wide mouth bottle with a narrower opening for controlled hydration.

  • Specially designed lip ideal for keeping the flow, without the stress of a mess
  • Tough and guaranteed for life
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 946 mL
Capacity (English) 32 oz.
Color Gray
Mouth Narrow
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.