missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 32 oz. Wide Mouth Bottle

Product Code. 17180056
Click to view available options
Color:
Blue
Gray
Seafoam
Trout Green
Unit Size:
Each
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 17180056

missing translation for 'mfr': Nalgene 21782024

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The Nalgene 32 oz. Wide Mouth water bottle is made with durable Tritan plastic, BPA/BPS-free, and made in the USA.

  • Expertly crafted threads for leak-proof hydration
  • Wide mouth that's ideal for ice cubes or fruit infusions
  • Proven in the elements and ready to withstand even the most rugged conditions
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 946 mL
Capacity (English) 32 oz.
Color Blue
Material Plastic
Mouth Wide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.