Learn More
Hamilton™ 33 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Product Code. 12311828
Change view
Click to view available options
Needle Gauge:
33 G
Unit Size:
Pack of 6
Product Code. Needle Gauge Unit Size
12311828 33 G Pack of 6
1 options
Product Code. 12311828 Supplier Hamilton™ Supplier No. 2203301
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Specification Sheet
  • Gauge – 33 gauge
  • Point Style – 2, 3, or 4
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – No Hub
Specifications
Needle Gauge 33 G
Product Type Hubless Needles