Learn More
Beckman Coulter Life Sciences 3400 Series High-Pressure Diffuser
Product Code. 31777212 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Connection Type:
3/8 in. Barb
Unit Size:
Each
Product Code. Connection Type Unit Size
31777212 3/8 in. Barb Each
1 options
Product Code. 31777212 Supplier Beckman Coulter Life Sciences Supplier No. 208073212
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Part of the MET ONE Facility Monitoring System.
  • 3400 Series High Pressure Diffuser for pressurized gas line testing
  • Flow rate 50 L/min., 1.77 CFM
  • 3/8 in. barb connection
Specifications
Product Type High-Pressure Diffuser
Display Type None
Connection Type 3/8 in. Barb
For Use With (Equipment) Beckman Coulter Analyzer