Learn More
STEMCELL Technologies 35 mm Culture Dishes
Product Code. p-7217426
Encompass_Preferred
Click to view available options
Quantity:
10 Dishes
500 Dishes
Unit Size:
Pack of 10
Pack of 500
Product Code. 15497311
Brand: STEMCELL Technologies 27100
Culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays
  • 35 mm culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays
  • They are manufactured from optical quality, thermally resistant, non-pyrogenic, non-toxic polystyrene
  • Recommended for: Hematopoietic progenitor cultures in methylcellulose-based media (MethoCult™)
  • Embryoid body formation from mouse embryonic and induced pluripotent stem cells
  • Performing the neural colony-forming cell assay (NCFC)
Specifications
For Use With (Application) Cell Culture; Colony Assay
Quantity 10 Dishes