missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequencing Standard, BigDye™ Terminator v1.1, for 3500/SeqStudio™ Flex

Product Code. 12010860 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12010860 4 Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12010860 Supplier Applied Biosystems™ Supplier No. 4404314
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye® Terminator v1.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.

The DNA contained in the preparation has been lyophilized to maximize stability.The two hydrolytic enzymes used in this kit, shrimp alkaline phosphatase and exonuclease I, effectively remove excess dNTPs and primers.

The BigDye Terminator v1.1 sequencing standard kit contains four tubes, each of which contains enough DNA of a known 1200 base pair sequence prepared with BigDye Terminator v1.1 to perform one spectral calibration⁄performance check and/or one control sequencing run on the Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The DNA contained in the preparation has been lyophilized to maximize stability.

Order Info

4 Tubes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye Terminator v1.1. Each tube can perform one spectral calibration⁄performance check and⁄or one control sequencing run on Applied Biosystems 3500 or SeqStudio Flex series genetic analyzers.

Store kit at -15 to -25°C.
Sample Type DNA
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye Terminator, BigDye
Product Type Sequencing Standards
Quantity 4 Tubes
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.