missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500xL Dx Capillary Array, 36 cm

Product Code. 15948826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15948826 1 Array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15948826 Supplier Applied Biosystems™ Supplier No. 4404686
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These internally uncoated 36-cm capillaries are supplied in assemblies of 24 capillaries per array, with a built-in frame for easy installation.

These internally uncoated 36-cm capillaries are supplied in assemblies of 24 capillaries per array with a built-in frame for easy installation. A radio-frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections.

This product is intended for in-vitro diagnostic use and is designed for use with the 3500 Dx and 3500xl Dx genetic analyzers and is not validated for use with other systems or applications.

The 3500 Dx and 3500xl Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

Note: The consumables are for distribution and use in specific European countries only. Not for use in the USA.

  • These internally uncoated 36 cm capillaries are supplied in preassembled sets assemblies of 24 capillaries per array, with a built-in frame for easy installation.
  • Radio frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The arrays may be used for 160 injections.
  • This product is intended for in-vitro diagnostic use on the Applied Biosystems 3500 Dx Series Genetic Analyzers and is not validated for use with other systems or applications.
  • The 3500 Dx Series Genetic Analyzer is to be used by trained operators in the analysis of human DNA or RNA, for the detection of genetic changes that may lead to disease presence or susceptibility.
  • Off-label uses such as fragment analysis are for Research Use Only.
  • The 3500 Dx Series Systems and consumables are for distribution and use in specific European countries only. Not for use in the USA.
TRUSTED_SUSTAINABILITY

Specifications

Starting Material DNA, RNA
Capillary Length 36 cm
Content And Storage Ambient, 1 array with 24 capillaries
For Use With (Equipment) 3500xL Dx Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.