missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Tubes™ 3810X
GREENER_CHOICE

Product Code. 10758052 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Eppendorf Quality
PCR Clean
Unit Size:
Pack of 1000

Product Code. 10758052

Brand: Eppendorf™ 0030125185

Only null left
Add to basket
Edge
Add to basket

The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.

Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.

  • Tube lid is easy to open and close for ergonomic operation.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • Graduation for visual control of the pipetted volume.
  • Frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86°C to 100°C.
  • Autoclavable when open (121°C, 20 min).
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade.
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Green
Material Polypropylene
For Use With (Application) Easy-to-open, securely closing, Eppendorf Tubes 3810X offers comfort and dependability for sample preparation, centrifugation and storage
No. per Pack 1000 Pk.
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Flat Frost Lid
Fit for Purpose Certified Quality
Bottom Shape Conical
Max. RCF 30,000 x g
Shape Cylindrical
Product Type Microcentrifuge Tube
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.