missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Plate Septa, 384 well

Product Code. 15889746
Change view
Click to view available options
Quantity:
20 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15889746 20 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889746 Supplier Applied Biosystems™ Supplier No. 4315934
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each package contains 20 septa, sufficient for 7,680 samples

These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.

These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.

Use with:
384-Well Plate Base and 384-Well Plate Retainer

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 20 septa, sufficient for 7,680 samples.
Format 384-well Plate
For Use With (Equipment) 3100-Avant Genetic Analyzer
Product Type Septa Plate
Quantity 20 each
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.