missing translation for 'onlineSavingsMsg'
Learn More

3Alpha-Hydroxy Pravastatin Lactone, TRC

Product Code. 30407174
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407174 5 mg -
30394650 10 mg -
30402343 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407174 Supplier Toronto Research Chemicals Supplier No. TRCH9523005MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85798-96-5
Molecular Formula C23 H34 O6
Molecular Weight (g/mol) 406.51
Synonym Butanoic acid, 2-methyl-, (1S,6S,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)-, Butanoic acid, 2-methyl-, (1S,6S,7R,8S)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)- (9CI), SQ 31906 lactone, (2S)-2-Methylbutanoic acid (1S,6S,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, 3α-Hydroxy pravastatin lactone, 3α-Hydroxypravastatin lactone, 6α-Hydroxy-iso-compactin
IUPAC Name [(1S,6S,7R,8S,8aR)-6-hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1CC=CC2=C[C@@H](O)[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H34O6/c1-4-13(2)23(27)29-20-7-5-6-15-10-19(25)14(3)18(22(15)20)9-8-17-11-16(24)12-21(26)28-17/h5-6,10,13-14,16-20,22,24-25H,4,7-9,11-12H2,1-3H3/t13-,14+,16+,17+,18-,19+,20-,22-/m0/s1
Formula Weight 406.2355
Chemical Name or Material 3a-Hydroxy Pravastatin Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.