missing translation for 'onlineSavingsMsg'
Learn More

(3aR,7aS)-rel-Hexahydro-1,3-isobenzofurandione, TRC

Product Code. 30398549
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398549 25 g 25g
30401248 5 g -
30390438 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398549 Supplier Toronto Research Chemicals Supplier No. TRCH29403025G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13149-00-3
Molecular Formula C8 H10 O3
Molecular Weight (g/mol) 154.16
Synonym 1,3-Isobenzofurandione, hexahydro-, (3aR,7aS)-rel-, 1,2-Cyclohexanedicarboxylic anhydride, cis- (8CI), 1,3-Isobenzofurandione, hexahydro-, cis-, rel-(3aR,7aS)-Hexahydro-1,3-isobenzofurandione, cis-1,2-Cyclohexanedicarboxylic anhydride, cis-Cyclohexane-1,2-dicarboxylic acid anhydride, cis-Hexahydrophthalic anhydride, rel-(3aR,7aS)-Hexahydroisobenzofuran-1,3-dione
IUPAC Name (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)[C@H]2CCCC[C@@H]12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2/t5-,6+
Formula Weight 154.063
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic acid anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.