missing translation for 'onlineSavingsMsg'
Learn More

3BDO, MedChemExpress

Product Code. 16492638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492638 5 mg 5mg
16480439 10 mg 10mg
16404106 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492638 Supplier Medchem Express Supplier No. HYU00434
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3BDO is a new mTOR activator which can also inhibit autophagy.

3BDO is a new mTOR activator which can also inhibit autophagy.

Specifications

Chemical Name or Material 3BDO
CAS 890405-51-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H17NO5
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (305.50 mM) ∣Ethanol : 20 mg/mL (61.10 mM; Need ultrasonic)
SMILES O=C1OC(COC2=C([N+]([O-])=O)C=CC=C2)CC1CC3=CC=CC=C3
Molecular Weight (g/mol) 327.33
Formula Weight 327.33
Percent Purity 99.91%
Grade Research
Physical Form Solid-Liquid Mixture
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.