missing translation for 'onlineSavingsMsg'
Learn More

3M™ Protective Coverall 4540+, Hooded

Product Code. 12652565
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12652565 Medium Pack of 20
11903794 Large Pack of 20
11993784 Small Pack of 20
12662595 X-Large Pack of 20
12672595 2X-Large Pack of 20
11933794 3X-Large Pack of 20
11943794 4X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12652565 Supplier 3M™ Supplier No. 7100040087
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Coverall is made from a laminated premium bicomponent non-woven and features a breathable SMMMS triangular back panel for improved comfort. Soft and lightweight material provides an ideal barrier to hazardous particles and limited liquid splashes.

  • Ideal barrier against dry particles and certain liquid chemical splash (CE Type 5/6)
  • Soft and light, ultra low-linting laminated microporous material that drapes and hangs well over the body facilitating ease of movement
  • 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Medium
Gender Unisex
Material Polyethylene, Polyester/Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Bound
Sterility Non-sterile
Protection Class CE Category III, Type 5/6 protection
Resistance Anti-static, Dust, Liquid
Coating Anti-static
Back Style Breathable SMMMS material
Certifications/Compliance CE, EN 13034, EN-ISO 13982-23, EN 1149-1:2006
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description White Polypropylene/Polyethylene laminated suit, SMMMS Polypropylene blue back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread
Latex Free No
Model 4540+
Size (English) Chest 36 to 39 in.
Size (English) Inseam 30 in.
Size (English) Waist 50 in.
Size (Metric) Chest 92 to 100 cm
Size (Metric) Inseam 76.2 cm
Size (Metric) Waist 127 cm
For Use With (Application) Pesticide application, asbestos and lead abatement, Coal dust in power plants, metal polishing, light-duty building Cleaning, machine or vehicle maintenance, paint spraying, pharmaceutical, general industrial clean-up, insulation laying, general powder handling and food processing
Height (English) 5.5 to 5.75 ft.
Height (Metric) 167 to 176 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.