missing translation for 'onlineSavingsMsg'
Learn More

3M™ Ethylene Oxide Monitor

Product Code. 15739767 Shop All 3M Products
Change view
Click to view available options
Product Type:
Ethylene Oxide Monitor only
Ethylene Oxide Monitor with Prepaid analysis
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15739767 Ethylene Oxide Monitor only Pack of 5
15739637 Ethylene Oxide Monitor with Prepaid analysis Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15739767 Supplier 3M™ Supplier No. 7000051749

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

No batteries, hoses, pumps or equipment to calibrate

  • Ideal for chemical, medical, and pharmaceutical labs.
  • Sensitivity of 0.24ppm-hours
  • Upper exposure limit of 600ppm-hours
  • Available as a monitor only or with prepaid analysis
  • These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
  • 3M Catalog# 3551

Misuse may result in sickness or death.

Compliance

Meets OSHA accuracy requirements.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Ideal for Chemical, Medical, and Pharmaceutical Labs
Certifications/Compliance OSHA
Detectable Analytes Ethylene Oxide
Measurement Range 600 ppm.Hour
Product Type Ethylene Oxide Monitor only
Model 3551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.