missing translation for 'onlineSavingsMsg'
Learn More

3M™ Standard Chemical Sorbents

Product Code. 10685315
Change view
Click to view available options
Capacity (English):
8 gal.
12 gal.
14 gal.
17 gal.
Type:
Chemical Sorbent Mini Boom
Chemical Sorbent Pad
Chemical Sorbent Particulate
Chemical Sorbent Pillow
Model:
P110
P200
P300
P500
Unit Size:
Each
Pack of 12
Pack of 16
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Type Model Unit Size
10685315 17 gal. Chemical Sorbent Pad P110 Pack of 200
12342459 12 gal. Chemical Sorbent Mini Boom P200 Pack of 12
12332459 8 gal. Chemical Sorbent Pillow P300 Pack of 16
12362459 14 gal. Chemical Sorbent Particulate P500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10685315 Supplier 3M™ Supplier No. 7000088220
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Absorb virtually any liquid with these chemical sorbents

Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.

  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA 29 CFR 1910.132
Color Yellow
Capacity (English) 17 gal.
Capacity (Metric) 64.3 L
Material Polypropylene
Type Chemical Sorbent Pad
For Use With (Application) For control of spills of certain hazardous liquids
Model P110
Dimensions (L x W) 33.02 x 27.94 cm (13 x 11 in.)
Length (English) 13 in.
Length (Metric) 33.02 cm
Width (English) 11 in.
Width (Metric) 27.94 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.