missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-3-Acetamidopyrrolidine, 98%, Thermo Scientific™

Product Code. 11363236
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363236 250 mg 250mg
11343236 1 g 1g
11353236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363236 Supplier Thermo Scientific Alfa Aesar Supplier No. L12567.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 131900-62-4
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00082729
InChI Key HDCCJUCOIKLZNM-ZCFIWIBFSA-N
Synonym 3r-+-3-acetamidopyrrolidine, r-n-pyrrolidin-3-yl acetamide, r-3-acetamidopyrrolidine, n-3r-pyrrolidin-3-yl acetamide, acetamide, n-3r-3-pyrrolidinyl, 3r---3-acetamidopyrrolidine, 3r-3-acetamidopyrrolidine, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, r-3-acetamido-pyrrolidine
PubChem CID 7016129
IUPAC Name N-[(3R)-pyrrolidin-3-yl]acetamide
SMILES CC(=O)NC1CCNC1

Specifications

Melting Point 52°C to 57°C
Boiling Point 70°C to 72°C (40 mmHg)
Quantity 250 mg
UN Number UN3259
Beilstein 4904138
Optical Rotation +46.5° (c=1 in Ethanol)
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material (3R)-(+)-3-Acetamidopyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.