missing translation for 'onlineSavingsMsg'
Learn More

(3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one, 98%, Thermo Scientific™

Product Code. 11486300
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486300 5 g 5g
11476300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486300 Supplier Thermo Scientific Alfa Aesar Supplier No. H27164.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97538-67-5
Molecular Formula C7H8Cl3NO2
Molecular Weight (g/mol) 244.496
MDL Number MFCD08274464
InChI Key GWQBXRYSVSZLSL-UJURSFKZSA-N
Synonym 3r,7as-3-trichloromethyl tetrahydropyrrolo 1,2-c oxazol-1 3h-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-hexahydropyrrolo 1,2-c 1,3 oxazol-1-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3,3,0 octane-4-one, 3r,7as-3-trichloromethyl-tetrahydro-3h-pyrrolo 1,2-c 1,3 oxazol-1-one, l-proline-chloral condensation product, 3r-trichloromethyl-cis-tetrahydropyrrolo 1,2-c oxazol-1-one, 2r,5s-2-trichloromethyl-1-aza-3-oxabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-tetrahydro-pyrrolo 1,2-c oxazol-1-one, 3r,7as-3-trichloromethyl-tetrahydropyrrolo 1,2-c oxazol-1 3h-one
PubChem CID 10944680
IUPAC Name (3R,7aS)-3-(trichloromethyl)-5,6,7,7a-tetrahydro-3H-pyrrolo[1,2-c][1,3]oxazol-1-one
SMILES C1CC2C(=O)OC(N2C1)C(Cl)(Cl)Cl

Specifications

Melting Point 109°C (decomposition)
Quantity 5 g
Optical Rotation −2.9° (c= 1 in Chloroform)
Formula Weight 244.51
Percent Purity 98%
Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.