missing translation for 'onlineSavingsMsg'
Learn More

(3S)-3-[4-[(2-Chloro-phenyl)methyl]phenoxy]tetrahydrofuran, TRC

Product Code. 30358600
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358600 50 mg 50mg
30353725 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358600 Supplier Toronto Research Chemicals Supplier No. TRCC35060850MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2517968-35-1
Molecular Formula C17H17ClO2
Molecular Weight (g/mol) 288.77
Synonym (S)-3-(4-(2-Chlorobenzyl)phenoxy)tetrahydrofuran
IUPAC Name (3S)-3-[4-[(2-chlorophenyl)methyl]phenoxy]oxolane
SMILES Clc1ccccc1Cc2ccc(O[C@H]3CCOC3)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H17ClO2/c18-17-4-2-1-3-14(17)11-13-5-7-15(8-6-13)20-16-9-10-19-12-16/h1-8,16H,9-12H2/t16-/m0/s1
Chemical Name or Material (3S)-3-[4-[(2-Chloro-phenyl)methyl]phenoxy]tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.