missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperazinyl)aniline, 95%

Product Code. 15441398
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441398 5 g 5g
15451398 250 mg 250mg
15431398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441398 Supplier Thermo Scientific Alfa Aesar Supplier No. H55224.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67455-41-8 | C10H15N3 | 177.25 g/mol

4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Reactive with oxidizing agents. Keep at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67455-41-8
Molecular Formula C10H15N3
Molecular Weight (g/mol) 177.25
MDL Number MFCD01365906
InChI Key VAVOYRCCWLRTMS-UHFFFAOYSA-N
Synonym 4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine
PubChem CID 422925
IUPAC Name 4-piperazin-1-ylaniline
SMILES NC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 119°C to 123°C
Color Purple to White
Quantity 5 g
Formula Weight 177.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(1-Piperazinyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.