missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperidinyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 15407718
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15407718 250 mg 250mg
15497708 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15407718

Brand: Thermo Scientific Alfa Aesar H61206.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1204-85-9
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD02575368
InChI Key ZEPXHFFGXQFUDP-UHFFFAOYSA-N
Synonym 4-piperidin-1-yl benzonitrile, 4-1-piperidinyl benzonitrile, 4-piperidin-1-yl-benzonitrile, 4-piperidinobenzonitrile, benzonitrile,4-1-piperidinyl, 4-piperidylbenzenecarbonitrile, acmc-209a7n, 4-piperidin-1-ylbenzenecarbonitrile
PubChem CID 766771
IUPAC Name 4-piperidin-1-ylbenzonitrile
SMILES C1CCN(CC1)C2=CC=C(C=C2)C#N

Specifications

Color White
Quantity 250 mg
UN Number UN3439
Formula Weight 186.26
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(1-Piperidinyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.