missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1-Pyrrolidinyl)benzonitrile, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Specifications
Specifications
| CAS Min % | 89.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 90% |
| Packaging | Amber glass bottle |
| Quantity | 10 g |
| Formula Weight | 172.23 |
| Percent Purity | 90% |
| Chemical Name or Material | 4-(1-Pyrrolidinyl)benzonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction