missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzonitrile, ≥90%, Thermo Scientific™

Product Code. 10311233
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10311233 1 g 1g
10036332 10 g 10g
10622495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10311233 Supplier Thermo Scientific Maybridge Supplier No. CC30116DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 10282-30-1
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD07368512
InChI Key ZNMSYUCZLWETII-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
PubChem CID 4961271
IUPAC Name 4-pyrrolidin-1-ylbenzonitrile
SMILES C1CCN(C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 90%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.