missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 96%

Product Code. 15449947
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449947 1 g 1g
15459947 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449947 Supplier Thermo Scientific Alfa Aesar Supplier No. H53186.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 389621-81-2 | C11H14BNO3 | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389621-81-2
Molecular Formula C11H14BNO3
Molecular Weight (g/mol) 219.05
MDL Number MFCD03411950
InChI Key VKPBESPVHGDDJS-UHFFFAOYSA-N
Synonym 4-pyrrolidine-1-carbonyl phenylboronic acid, 4-pyrrolidine-1-carbonyl phenyl boronic acid, 4-pyrrolidinylcarbonylphenylboronic acid, 4-pyrrolidine-1-carbonyl benzeneboronic acid, 4-boronophenyl pyrrolidin-1-yl methanone, 1-4-boronobenzoyl pyrrolidine, boronic acid, 4-1-pyrrolidinylcarbonyl phenyl, 4-borono-n-1-pyrrolidinyl benzamide, 4-pyrrolidin-1-ylcarbonyl phenyl boronic acid
PubChem CID 2773571
IUPAC Name [4-(pyrrolidine-1-carbonyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(=O)N1CCCC1

Specifications

Melting Point 136°C to 140°C
Quantity 1 g
Formula Weight 219.05
Percent Purity 96%
Chemical Name or Material 4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.