missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolyl)benzoic acid, 99%

Product Code. 15427957
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427957 Supplier Thermo Scientific Alfa Aesar Supplier No. H51073.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22106-33-8 | C11H8NO2 | 186.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22106-33-8
Molecular Formula C11H8NO2
MDL Number MFCD00082569
Quantity 1 g
Sensitivity Moisture sensitive
Synonym 4-1h-pyrrol-1-yl benzoic acid, 4-1-pyrrolyl benzoic acid, benzoic acid, 4-1h-pyrrol-1-yl, n-4-carboxy phenylpyrrole, 4-pyrrol-1-yl benzoic acid, 4-pyrrolylbenzoic acid, maybridge1_008067, 4-1-pyrrolo benzoic acid, 4-pyrrol-1-yl-benzoic acid, 4-1h-pyrrol-1yl benzoic acid
InChI Key NLSIIPKSANRIGS-UHFFFAOYSA-M
SMILES [O-]C(=O)C1=CC=C(C=C1)N1C=CC=C1
IUPAC Name 4-(1H-pyrrol-1-yl)benzoate
Molecular Weight (g/mol) 186.19
PubChem CID 2737256
Formula Weight 187.2
Percent Purity 99%
Chemical Name or Material 4-(1-Pyrrolyl)benzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.