missing translation for 'onlineSavingsMsg'
Learn More

4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid, ≥90%, Thermo Scientific™

Product Code. 10593393
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10593393 250 mg 250mg
10289492 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10593393 Supplier Thermo Scientific Maybridge Supplier No. MO00953CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 162046-56-2
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD06658985
InChI Key LUVXDMFUUZGFST-UHFFFAOYSA-N
Synonym 4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid
PubChem CID 2794649
IUPAC Name 4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 321.37
Percent Purity 97%
Chemical Name or Material 4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.