missing translation for 'onlineSavingsMsg'
Learn More

4-{[1-(tert-butoxycarbonyl)piperid-4-yloxy]methyl}benzoic acid, 97%, Thermo Scientific™

Product Code. 10799415
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10799415 5 g Amber Glass Bottle 5g
10134154 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10799415 Supplier Thermo Scientific Maybridge Supplier No. CC67901DE
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-52-2
Molecular Formula C18H25NO5
Molecular Weight (g/mol) 335.4
MDL Number MFCD11841069
InChI Key CGLNCSVUPUKDKT-UHFFFAOYSA-N
Synonym 4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzoic acid, 4-1-tert-butoxycarbonyl piperid-4-yloxy methyl benzoic acid, 4-4-carboxybenzyl oxy piperidine, n-boc protected
PubChem CID 43811046
IUPAC Name 4-[[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxymethyl]benzoic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 335.4
Percent Purity 97%
Chemical Name or Material 4-{[1-(Tert-butoxycarbonyl)piperid-4-yloxy]methyl}benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.