missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Imidazol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10121162
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121162 250 mg 250mg
10572842 1 g 1g
10505675 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10121162 Supplier Thermo Scientific Maybridge Supplier No. CC18509CB
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 86718-08-3
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD02682059
InChI Key SRQXVPAXMMIUFH-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-yl phenyl methanol, 4-imidazol-1-yl-phenyl methanol, 4-1-imidazolyl phenyl methanol, 4-imidazol-1-ylphenyl methanol, 4-1-imidazolyl benzyl alcohol, 4-imidazol-1-yl phenyl methanol, benzenemethanol, 4-1h-imidazol-1-yl, acmc-209qb5, 4-imidazolylphenyl methan-1-ol
PubChem CID 2776486
IUPAC Name (4-imidazol-1-ylphenyl)methanol
SMILES C1=CC(=CC=C1CO)N2C=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material [4-(1H-Imidazol-1-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.