missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)phenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10435312
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10435312 250 mg Amber Glass Bottle 250mg
10364372 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10435312 Supplier Thermo Scientific Maybridge Supplier No. CC18407CB
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 352018-96-3
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.25
MDL Number MFCD02681939
InChI Key GLAOJNMWLIFKCG-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl phenyl isothiocyanate, 1-4-isothiocyanatophenyl pyrazole, 4-1h-pyrazol-1-yl phenylisothiocyanate, 1h-pyrazole,1-4-isothiocyanatophenyl, 1h-pyrazole, 1-4-isothiocyanatophenyl, 4-pyrazolylbenzenisothiocyanate, 4-1h-pyrazole-1-yl phenyl isothiocyanate
PubChem CID 2776478
IUPAC Name 1-(4-isothiocyanatophenyl)pyrazole
SMILES S=C=NC1=CC=C(C=C1)N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.25
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.