missing translation for 'onlineSavingsMsg'
Learn More

4-[[2-[2-[(6-Chlorohexyl)oxy]ethoxy]ethyl]amino]-4-oxo-butanoic Acid, TRC

Product Code. 30358240
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358240 10 mg -
30358133 100 mg 100mg
30358131 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358240 Supplier Toronto Research Chemicals Supplier No. TRCC36854310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1488363-39-8
Molecular Formula C14H26ClNO5
Molecular Weight (g/mol) 323.81
IUPAC Name 4-[2-[2-(6-chlorohexoxy)ethoxy]ethylamino]-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)NCCOCCOCCCCCCCl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H26ClNO5/c15-7-3-1-2-4-9-20-11-12-21-10-8-16-13(17)5-6-14(18)19/h1-12H2,(H,16,17)(H,18,19)
Formula Weight 323.15
Chemical Name or Material 4-((2-(2-((6-Chlorohexyl)oxy)ethoxy)ethyl)amino)-4-oxo-butanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.