missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3-Epoxypropoxy)phenylacetamide, TRC

Product Code. 30378133
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378133 250 mg
30374284 100 mg
30378377 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378133 Supplier Toronto Research Chemicals Supplier No. TRCE589600250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29122-69-8
Molecular Formula C11 H13 N O3
Molecular Weight (g/mol) 207.23
Synonym Benzeneacetamide, 4-(2-oxiranylmethoxy)-, Acetamide, 2-[p-(2,3-epoxypropoxy)phenyl]- (8CI), Benzeneacetamide, 4-(oxiranylmethoxy)- (9CI), 4-(2-Oxiranylmethoxy)benzeneacetamide, 1-(p-Carbamoylmethylphenoxy)-2,3-epoxypropane, 2-[4-(Oxiran-2-ylmethoxy)phenyl]acetamide, 4-(2,3-Epoxypropoxy)phenylacetamide, 4-(2,3-Oxidopropoxy)phenylacetamide, 2-[4-[[(2RS)-Oxiran-2-yl]methoxy]phenyl]acetamide
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)phenyl]acetamide
SMILES NC(=O)Cc1ccc(OCC2CO2)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H13NO3/c12-11(13)5-8-1-3-9(4-2-8)14-6-10-7-15-10/h1-4,10H,5-7H2,(H2,12,13)
Formula Weight 207.0895
Percent Purity >95%
Chemical Name or Material 2-[4-[[(2RS)-Oxiran-2-yl]methoxy]phenyl]acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.