missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline, 97%

Product Code. 15400836
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400836 1 g 1g
15420836 250 mg 250mg
15410836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400836 Supplier Thermo Scientific Alfa Aesar Supplier No. H54156.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 82925-02-8 | C19H24N2O2 | 312.41 g/mol

It is an intermediate for the preparation of Elacridar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for the preparation of Elacridar.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82925-02-8
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.41
MDL Number MFCD04974539
InChI Key DGOOLMGPMIHRFY-UHFFFAOYSA-N
Synonym 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl aniline, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydro-2-isoquinolinyl ethyl aniline, 4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl-benzenamine, 4-2-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl ethyl aniline, benzenamine,4-2-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl ethyl, 4-2-6,7-dimethoxy-3,4-dihydroisoquinoline-2 1h-yl-ethyl-benzenamine, 4-2-1,2,3,4-tetrahydro-6,7-dimethoxy-2-isoquinolyl ethyl benzenamine, 4-2-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-yl ethyl benzenamine, 4-2-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl ethyl benzenamine
PubChem CID 10041227
IUPAC Name 4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
SMILES COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1

Specifications

Quantity 1 g
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 312.41
Percent Purity 97%
Chemical Name or Material 4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.