missing translation for 'onlineSavingsMsg'
Learn More

AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents

Product Code. 12194471
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
Unit Size:
100mg
10mg
1g
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12194471 100 mg Amber Glass 100mg
10357924 10 mg Amber Glass 10mg
10707735 50 mg Amber Glass 50mg
10104144 500 mg Amber Glass 500mg
11747499 1 g Amber Glass 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12194471 Supplier Fisher BioReagents Supplier No. BP2644100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Irreverisble serine protease inhibitor
  • Inhibits proteases like trypsin, chymotrypsin, kallikrein, plasmin, and thrombin
  • Acts as Sulfonyl fluoride that reacts with hydroxy group of the active site serine residue
  • More soluble and less toxic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Molecular Weight (g/mol) 239.689
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

Melting Point 175°C
Color White
Infrared Spectrum Pass Test
Assay Percent Range ≥97 %
Packaging Amber Glass
Quantity 100 mg
Formula Weight 239.7
Percent Purity 97% min.
Physical Form Powder or Solid
Chemical Name or Material 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.