missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals

Product Code. 11414822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414822 100 mg 100mg
11424822 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414822 Supplier MP Biomedicals Supplier No. 0219350380
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Molecular Weight (g/mol) 239.689
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

Quantity 100 mg
Formula Weight 239.7
Chemical Name or Material 4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride
compliance-icons
Product Identifier
  • 4-(2-Aminoethyl)benzenesulfonyl Fluoride Hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4-(2-Aminoethyl)benzenesulfonyl FluorideHydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.