missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals™

Product Code. 11434822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434822 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434822 Supplier MP Biomedicals Supplier No. 0219350391
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers many advantages compared to PMSF and DFP. 4-(2-Aminoethyl)Benzenesulfonyl Fluoride Hydrochloride, MP Biomedicals is a covalent binding serine protease inhibitor. AEBSF belongs to the family of irreversible sulfonyl fluoride inhibitors that block tr

AEBSF is an irreversible serine protease inhibitor that has been shown to inhibit trypsin, chymotrypsin, plasmin kallikrein and thrombin. It is well suited for use in tissue culture media as well as in large scale purification of recombinant proteins, susceptible to proteolysis.

AEBSF is an alternative to PMSF, DFP and related organophosphate inhibitors.

  • Similar in structure, with similar inhibition constants
  • Lower molecular weight and toxicity
  • Offers better solubility in water, lower toxicity, greater stability and higher inhibitory activity

Specifications

CAS 30827-99-7
Melting Point 175°C to 185°C
Molecular Formula C8H11ClFNO2S
Quantity 1 g
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
Solubility Information Soluble in methanol (10mg/mL, clear, colorless solution).
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
Molecular Weight (g/mol) 239.689
PubChem CID 186136
Formula Weight 239.7
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) Working Concentration: AEBSF has been used in cell culture in concentrations of up to 0.25mM.
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.